missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%

Manufacturer:  Alfa Aesar™ L09558MD

 View more versions of this product

Catalog No. AAL09558MD



Description

Description

Applications
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine is used as chiral solvation agent for determination of enantiomeric excess of chiral acids by NMR. It is used in various catalyst systems for asymmetric reactions.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat, air and oxidizing agents.
Specifications

Specifications

(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine
80°C to 84°C
35132-20-8
MFCD00082769
2369988
PONXTPCRRASWKW-ZIAGYGMSSA-N
(1R,2R)-1,2-diphenylethane-1,2-diamine
2724998
≥98%
+104° (c=1 in Methanol)
250mg
C14H16N2
UN3259
1r,2r-+-1,2-diphenylethylenediamine, 1r,2r-1,2-diphenylethane-1,2-diamine, 1r,2r-rel-1,2-diphenylethane-1,2-diamine, 1r,2r-1,2-diphenylethylenediamine, 1r,2r-+-1,2-diamino-1,2-diphenylethane, 1r,2r-1,2-diphenyl-1,2-ethanediamine, 1r,2r-+-1,2-diphenyl-1,2-ethanediamine, +-1,2-diphenylethylenediamine, +-stilbenediamine, r,r-dpen
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N
212.296
212.3

Chemical Identifiers

35132-20-8
212.296
PONXTPCRRASWKW-ZIAGYGMSSA-N
2724998
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N
C14H16N2
MFCD00082769
1r,2r-+-1,2-diphenylethylenediamine, 1r,2r-1,2-diphenylethane-1,2-diamine, 1r,2r-rel-1,2-diphenylethane-1,2-diamine, 1r,2r-1,2-diphenylethylenediamine, 1r,2r-+-1,2-diamino-1,2-diphenylethane, 1r,2r-1,2-diphenyl-1,2-ethanediamine, 1r,2r-+-1,2-diphenyl-1,2-ethanediamine, +-1,2-diphenylethylenediamine, +-stilbenediamine, r,r-dpen
(1R,2R)-1,2-diphenylethane-1,2-diamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

Recommended Storage : 2° to 8°C

Documents