Please login to your online account to display your discounted pricing

(1R,2R)-(-)-1,2-Diaminocyclohexane, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC364640050

 View more versions of this product

Catalog No. AC364640050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Low Melting Solid
Formula Weight 114.19
Quantity 5g
Color White to pale yellow
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific Rotation Condition − 24.50 (26.50°C c=5 1 N HCl 20°)
Melting Point 40° to 45°C
Beilstein 13, III, 6
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C6H14N2
CAS 20439-47-8
MDL Number MFCD00062985
Health Hazard 1 Danger
CAS Min % 98.5
Packaging Glass bottle
Specific Rotation -24.5° to -26.5° (20°C, 589nm) (c=5, 1N HCl)
Linear Formula C6H10(NH2)2
Chemical Name or Material (1R, 2R)-(-)-1, 2-Diaminocyclohexane
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

Specifications

Chemical Name or Material (1R, 2R)-(-)-1, 2-Diaminocyclohexane
CAS 20439-47-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H14N2
Linear Formula C6H10(NH2)2
Formula Weight 114.19
MDL Number MFCD00062985
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

SDS & Safety Documents