missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2R)-(-)-1,2-Diaminocyclohexane, 98%

Manufacturer:  Alfa Aesar™ L1405906

 View more versions of this product

Catalog No. AAL1405906



Description

Description

Applications
(1R,2R)-(-)-1,2-Diaminocyclohexane is a versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
Specifications

Specifications

(1R,2R)-(-)-1,2-Diaminocyclohexane
Amine-like
Air sensitive
40°C to 43°C
20439-47-8
MFCD00062985
4780911
Soluble in water.
C1CCC(C(C1)N)N
114.192
114.19
70°C (158°F)
−25.5° (c=5 in 1N HCl)
104°C to 110°C (40mmHg)
5g
C6H14N2
UN3259
1r,2r---1,2-cyclohexanediamine, 1r,2r---1,2-diaminocyclohexane, 1r,2r-1,2-cyclohexanediamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, 1r,2r-diaminocyclohexane, r-dach, trans-1,2-cyclohexanediamine, trans-1,2-diaminocyclohexane, trans-cyclohexane-1,2-diamine
SSJXIUAHEKJCMH-PHDIDXHHSA-N
(1R,2R)-cyclohexane-1,2-diamine
43806
98%

Chemical Identifiers

20439-47-8
114.192
SSJXIUAHEKJCMH-PHDIDXHHSA-N
43806
C1CCC(C(C1)N)N
C6H14N2
MFCD00062985
1r,2r---1,2-cyclohexanediamine, 1r,2r---1,2-diaminocyclohexane, 1r,2r-1,2-cyclohexanediamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, 1r,2r-diaminocyclohexane, r-dach, trans-1,2-cyclohexanediamine, trans-1,2-diaminocyclohexane, trans-cyclohexane-1,2-diamine
(1R,2R)-cyclohexane-1,2-diamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P261-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P333+P313-P363-P501c

H314-H317-H318-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Keep cold

Documents