Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R)-10-Camphorsulfonamide, 97%

$56.20 - $174.78

Chemical Identifiers

Chemical Name or Material (1R)-10-Camphorsulfonamide
CAS 72597-34-3
Molecular Formula C10H17NO3S
Formula Weight 231.32
MDL Number MFCD00151500
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4435803

  SDS
alfa aesar™
4435803
1g Each for $56.20

AA4435806

  SDS
alfa aesar™
4435806
5g Each for $174.78
Description & Specifications

Specifications

Formula Weight 231.32
Melting Point 134°C (dec.)
Physical Form Powder
Quantity 1g
Assay 97%
MDL Number MFCD00151500
CAS 72597-34-3
Molecular Formula C10H17NO3S
Chemical Name or Material (1R)-10-Camphorsulfonamide