missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R)-10-Camphorsulfonamide, 97%

Manufacturer:  Alfa Aesar™ 4435806

 View more versions of this product

Catalog No. AA4435806



Specifications

Specifications

(1R)-10-Camphorsulfonamide
5g
C10H17NO3S
1r---10-camphorsulfonamide
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)N)C
231.31
231.32
97%
134°C (decomposition)
72597-34-3
MFCD00151500
SBLUNABTQYDFJM-MHPPCMCBSA-N
[(4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonamide
12840037
Powder

Chemical Identifiers

72597-34-3
231.31
SBLUNABTQYDFJM-MHPPCMCBSA-N
12840037
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)N)C
C10H17NO3S
MFCD00151500
1r---10-camphorsulfonamide
[(4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents