Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt, 97%

Chemical Identifiers

Chemical Name or Material 1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt
CAS 96107-94-7
Molecular Formula C4H5N4NaO2
Formula Weight 164.11
MDL Number MFCD02683105
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2346106

  SDS
alfa aesar
B2346106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2346114

  SDS
alfa aesar™
B2346114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2346122

  SDS
alfa aesar™
B2346122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Health Hazard 2 GHS H Statement
H228
Formula Weight 164.11
Quantity 5g
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
UN Number UN1325
MDL Number MFCD02683105
Assay 97%
Health Hazard 1 Warning
Chemical Name or Material 1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt
Assay Percent Range 0.99
Molecular Formula C4H5N4NaO2
CAS 96107-94-7
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a

GHS H Statement
H228