missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H-Tetrazole-5-acetic Acid 98.0+%, TCI America™
Supplier: TCI America T27375G
Specifications
1H-Tetrazole-5-acetic Acid | |
177°C | |
C3H4N4O2 | |
5 g | |
JUNAPQMUUHSYOV-UHFFFAOYSA-N | |
2-(2H-tetrazol-5-yl)acetic acid | |
89031 | |
≥98.0% (GC,T) |
21743-75-9 | |
White | |
MFCD03094010 | |
1325 | |
C(C1=NNN=N1)C(=O)O | |
128.091 | |
128.09 | |
Crystalline Powder |
Chemical Identifiers
21743-75-9 | |
128.091 | |
JUNAPQMUUHSYOV-UHFFFAOYSA-N | |
2-(2H-tetrazol-5-yl)acetic acid |
C3H4N4O2 | |
MFCD03094010 | |
89031 | |
C(C1=NNN=N1)C(=O)O |
Safety and Handling
TSCA : No