missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Pyrrolo[3,2-b]pyridine, TCI America™

Manufacturer:  TCI America P20415G

 View more versions of this product

Catalog No. P20415G



Specifications

Specifications

1H-Pyrrolo[3,2-b]pyridine
128°C
272-49-1
C7H6N2
4-Azaindole
C1=CC2=C(C=CN2)N=C1
118.139
118.14
Yellow
5g
≥98.0% (GC,T)
MFCD00971977
XWIYUCRMWCHYJR-UHFFFAOYSA-N
1H-pyrrolo[3,2-b]pyridine
9226
Crystal-Powder at 20°C

Chemical Identifiers

272-49-1
118.139
XWIYUCRMWCHYJR-UHFFFAOYSA-N
9226
C1=CC2=C(C=CN2)N=C1
C7H6N2
MFCD00971977
4-Azaindole
1H-pyrrolo[3,2-b]pyridine
Safety and Handling

Safety and Handling

RTECSNumber : UY8715000

TSCA : No

Documents