missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Pyrazole-1-carboxamidine monohydrochloride, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros AC368250500

 View more versions of this product

Catalog No. AC368250500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1H-Pyrazole-1-carboxamidine monohydrochloride
167° to 170°C
99%
98.5
Authentic
Crystals or Crystalline Powder
99%
146.58
1-Amidinopyrazole monohydrochloride
White to light yellow
50g
775445
100.0
Glass bottle
≥98.5%
C4H6N4·HCl
MFCD00210087

Chemical Identifiers

775445
100.0
C4H6N4·HCl
MFCD00210087
1H-Pyrazole-1-carboxamidine monohydrochloride, 99%
98.5
99%
146.58
1-Amidinopyrazole monohydrochloride
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.