Please login to your online account to display your discounted pricing

1H-Pyrazole-1-carboxamidine monohydrochloride, 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC368250100

 View more versions of this product

Catalog No. AC368250100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 146.58
Quantity 10g
Color White to light yellow
Melting Point 167° to 170°C
CAS 775445
CAS Max % 100.0
Chemical Name or Material 1H-Pyrazole-1-carboxamidine monohydrochloride
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C4H6N4·HCl
MDL Number MFCD00210087
Synonym 1-Amidinopyrazole monohydrochloride
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1H-Pyrazole-1-carboxamidine monohydrochloride
Name Note 99%
CAS 775445
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H6N4·HCl
Formula Weight 146.58
MDL Number MFCD00210087
Synonym 1-Amidinopyrazole monohydrochloride
Accessories

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.

SDS & Safety Documents