Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H-Perfluorohexane, 97%

$143.78 - $1,302.23

Chemical Identifiers

Chemical Name or Material 1H-Perfluorohexane
CAS 355-37-3
Molecular Formula C6HF13
Linear Formula CF3(CF2)5H
Formula Weight 320.05
MDL Number MFCD00142632
Beilstein 1799000
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2022606

  SDS
alfa aesar™
B2022606
5g Each for $143.78

AAB2022614

  SDS
alfa aesar™
B2022614
25g Each for $398.68

AAB2022622

  SDS
alfa aesar™
B2022622
100g Each for $1,302.23
Description & Specifications

Specifications

Formula Weight 320.05
Chemical Name or Material 1H-Perfluorohexane
Assay 97%
Molecular Formula C6HF13
Linear Formula CF3(CF2)5H
MDL Number MFCD00142632
CAS 355-37-3
Beilstein 1799000
Boiling Point 70° to 71°C
Quantity 5g