missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Indole-7-carboxylic acid, 90%, Maybridge

Chemical Identifiers

CAS 1670-83-3
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00210442
InChI Key IPDOBVFESNNYEE-UHFFFAOYSA-N
Synonym 1h-indole-7-carboxylicacid
PubChem CID 74281
IUPAC Name 1H-indole-7-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)NC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB12826CB SDS View Documents Fisher Scientific Company
BTB12826CB
250mg Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1670-83-3
161.16
IPDOBVFESNNYEE-UHFFFAOYSA-N
74281
C1=CC2=C(C(=C1)C(=O)O)NC=C2
C9H7NO2
MFCD00210442
1h-indole-7-carboxylicacid
1H-indole-7-carboxylic acid

Specifications

Amber Glass Bottle
1670-83-3
100.0
C9H7NO2
1h-indole-7-carboxylicacid
C1=CC2=C(C(=C1)C(=O)O)NC=C2
161.16
161.16
1H-Indole-7-carboxylic acid
250mg
89.5
90%
MFCD00210442
IPDOBVFESNNYEE-UHFFFAOYSA-N
1H-indole-7-carboxylic acid
74281
90%