Please login to your online account to display your discounted pricing

1H-Indole-3-carboxylic acid, 97%


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC12701DA

 View more versions of this product

Catalog No. CC12701DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161.16
Quantity 1g
Assay 97%
Chemical Name or Material 1H-Indole-3-carboxylic acid
CAS 771-50-6
Health Hazard 1 Warning
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H7NO2

Specifications

Chemical Name or Material 1H-Indole-3-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7NO2
Formula Weight 161.16
Accessories

Warning

SDS & Safety Documents