Please login to your online account to display your discounted pricing

1H-Indole-3-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 1H-Indole-3-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7NO2
Formula Weight 161.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC12701DA

  SDS
fisher scientific company
CC12701DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.16
Quantity 1g
Assay 97%
Chemical Name or Material 1H-Indole-3-carboxylic acid
CAS 771-50-6
Health Hazard 1 Warning
Packaging Amber Glass Bottle
Molecular Formula C9H7NO2
Assay Percent Range 97%
Safety and Handling

Warning