Please login to your online account to display your discounted pricing

1H-Indole-3-carbonitrile, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RF01264EA

 View more versions of this product

Catalog No. RF01264EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 142.16
Quantity 10g
Molecular Formula C9H6N2
Chemical Name or Material 1H-Indole-3-carbonitrile
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 5457-28-3

Specifications

Chemical Name or Material 1H-Indole-3-carbonitrile
Assay Percent Range 97%
Molecular Formula C9H6N2
Formula Weight 142.16