Please login to your online account to display your discounted pricing

1H-Indole-3-carbonitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1H-Indole-3-carbonitrile
Assay Percent Range 97%
Molecular Formula C9H6N2
Formula Weight 142.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF01264EA

 
fisher scientific company
RF01264EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.16
Quantity 10g
Molecular Formula C9H6N2
Chemical Name or Material 1H-Indole-3-carbonitrile
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 5457-28-3