missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Indole-3-acetic acid, 99+%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 122160100

 View more versions of this product

Catalog No. AC122160100



Description

Description

Specifications

Specifications

1H-Indole-3-acetic acid
Authentic
0.5% max. (K.F.)
167.0°C
87-51-4
100.0
C10H9NO2
22, 66
1h-indol-3-ylacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, 3-indoleacetic acid, heteroauxin, indol-3-ylacetic acid, indole-3-acetic acid, indoleacetic acid, rhizopin, rhizopon a
SEOVTRFCIGRIMH-UHFFFAOYSA-N
2-(1H-indol-3-yl)acetic acid
802
175.19
99+%
171°C
Glass bottle
Colorless to Yellow
10g
99.0
99% min. (HPLC)
MFCD00005636
15, 5004
Solubility in water: 8g/L (20°C). Other solubilities: soluble in acetone and ether,freely soluble in alcohol,sparingly soluble in chloroform
C1=CC=C2C(=C1)C(=CN2)CC(=O)O
175.19
CHEBI:16411
Liquid

Chemical Identifiers

87-51-4
175.19
SEOVTRFCIGRIMH-UHFFFAOYSA-N
802
2-(1H-indol-3-yl)acetic acid
C10H9NO2
MFCD00005636
1h-indol-3-ylacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, 3-indoleacetic acid, heteroauxin, indol-3-ylacetic acid, indole-3-acetic acid, indoleacetic acid, rhizopin, rhizopon a
CHEBI:16411
C1=CC=C2C(=C1)C(=CN2)CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 201-748-2

SDS Documents