missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H-Indole-3-acetic acid, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC122165000
Specifications
1H-Indole-3-acetic acid | |
Authentic | |
0.5% max. (K.F.) | |
167.0°C | |
87-51-4 | |
100.0 | |
C10H9NO2 | |
22, 66 | |
1h-indol-3-ylacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, 3-indoleacetic acid, heteroauxin, indol-3-ylacetic acid, indole-3-acetic acid, indoleacetic acid, rhizopin, rhizopon a | |
SEOVTRFCIGRIMH-UHFFFAOYSA-N | |
2-(1H-indol-3-yl)acetic acid | |
802 | |
175.19 | |
99+% |
171°C | |
Glass bottle | |
Colorless to Yellow | |
500g | |
99.0 | |
99% min. (HPLC) | |
MFCD00005636 | |
15, 5004 | |
Solubility in water: 8g/L (20°C). Other solubilities: soluble in acetone and ether,freely soluble in alcohol,sparingly soluble in chloroform | |
C1=CC=C2C(=C1)C(=CN2)CC(=O)O | |
175.19 | |
CHEBI:16411 | |
Liquid |
Chemical Identifiers
87-51-4 | |
175.19 | |
SEOVTRFCIGRIMH-UHFFFAOYSA-N | |
802 | |
2-(1H-indol-3-yl)acetic acid |
C10H9NO2 | |
MFCD00005636 | |
1h-indol-3-ylacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, 3-indoleacetic acid, heteroauxin, indol-3-ylacetic acid, indole-3-acetic acid, indoleacetic acid, rhizopin, rhizopon a | |
CHEBI:16411 | |
C1=CC=C2C(=C1)C(=CN2)CC(=O)O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 201-748-2