Please login to your online account to display your discounted pricing

1H-Indole-2-carboxylic acid, 97%


Quantity 5g

Manufacturer: fisher scientific company  CC12801DE

 View more versions of this product

Catalog No. CC12801DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161.16
Quantity 5g
Assay 97%
Chemical Name or Material 1H-Indole-2-carboxylic acid
Molecular Formula C9H7NO2
CAS 1477-50-5
Health Hazard 1 Warning
Assay Percent Range 97%

Specifications

Chemical Name or Material 1H-Indole-2-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7NO2
Formula Weight 161.16
Accessories

Warning

SDS & Safety Documents