Learn More
1H-Indene, 90+%, stab. with 0.01% 4-tert-butylcatechol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A12192AU
Description
1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.
Solubility
Soluble in ethanol, acetone, ethyl ether, and ether. Insoluble in water.
Notes
Store at 4°C. Store in dark. Incompatible with oxidizing agent, light, heat.
Specifications
1H-Indene | |
95-13-6 | |
0.996 | |
58°C (136°F) | |
C9H8 | |
MFCD00003777 | |
UN3295 | |
Light sensitive | |
indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy | |
YBYIRNPNPLQARY-UHFFFAOYSA-N | |
1H-indene | |
7219 | |
116.16 |
Stabilized with 0.01% 4-tert-butylcatechol | |
∼-2°C | |
181°C to 182°C | |
Aromatic | |
1.57 | |
1000 mL | |
635873 | |
14,4939 | |
Soluble in ethanol,acetone,ethyl ether,and ether. Insoluble in water. | |
C1C=CC2=CC=CC=C21 | |
116.163 | |
CHEBI:41921 | |
≥90% |
Chemical Identifiers
95-13-6 | |
116.163 | |
YBYIRNPNPLQARY-UHFFFAOYSA-N | |
7219 | |
1H-indene |
C9H8 | |
MFCD00003777 | |
indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy | |
CHEBI:41921 | |
C1C=CC2=CC=CC=C21 |
Safety and Handling
GHS H Statement
H304-H226-H332
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P303+P361+P353-P304+P340-P312-P314-P370+P378q-P501c
H226-H335-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 202-393-6
RTECSNumber : NK8225000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only