Please login to your online account to display your discounted pricing

1H-Indazole-7-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1H-Indazole-7-carboxylic acid
CAS 677304-69-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD06804572
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC429600010

  SDS
thermo fisher scientific
AC429600010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC429600050

  SDS
thermo fisher scientific
AC429600050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 1H-Indazole-7-carboxylic acid
CAS 677304-69-7
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD06804572
Melting Point 228° to 233°C
Color White to yellow
Quantity 1g
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.