missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H-Indazole-6-carboxaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 431040050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1H-Indazole-6-carboxaldehyde | |
96.0 | |
Authentic | |
Glass Bottle | |
5g | |
JTWYTTXTJFDYAG-UHFFFAOYSA-N | |
1H-indazole-6-carbaldehyde | |
18538368 | |
97% |
669050-69-5 | |
100.0 | |
96% min. (HPLC) | |
C8H6N2O | |
1h-indazole-6-carboxaldehyde, 6-formylindazole, 6-formyl-1h-indazole, 2h-indazole-6-carbaldehyde, indazole-6-carboxaldehyde, 1h-indazole-6-cabaldehyde, pubchem23878, acmc-1bc45, 2h-indazole-6-carboxaldehyde, 1-h-indazole-6-carboxaldehyde | |
C1=CC2=C(C=C1C=O)NN=C2 | |
146.15 | |
146.15 |
Chemical Identifiers
669050-69-5 | |
146.15 | |
1h-indazole-6-carboxaldehyde, 6-formylindazole, 6-formyl-1h-indazole, 2h-indazole-6-carbaldehyde, indazole-6-carboxaldehyde, 1h-indazole-6-cabaldehyde, pubchem23878, acmc-1bc45, 2h-indazole-6-carboxaldehyde, 1-h-indazole-6-carboxaldehyde | |
1H-indazole-6-carbaldehyde |
C8H6N2O | |
JTWYTTXTJFDYAG-UHFFFAOYSA-N | |
18538368 | |
C1=CC2=C(C=C1C=O)NN=C2 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Take off contaminated clothing and wash before reuse.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Warning
RUO â Research Use Only