Please login to your online account to display your discounted pricing

1H-Indazole-4-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1H-Indazole-4-carboxylic acid
CAS 677306-38-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD03840644
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC430090050

  SDS
thermo fisher scientific
AC430090050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC430090010

  SDS
thermo fisher scientific
AC430090010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 162.15
Quantity 5g
Color White to cream
Melting Point 207° to 212°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
CAS 677306-38-6
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material 1H-Indazole-4-carboxylic acid
Assay Percent Range 97%
Molecular Formula C8H6N2O2
MDL Number MFCD03840644
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.