Learn More
1H-Indazole-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6400114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1H-Indazole-3-carboxylic acid | |
266°C to 270°C (literature) | |
C8H6N2O2 | |
25 g | |
indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid | |
C1=CC=C2C(=C1)C(=NN2)C(=O)O | |
162.148 | |
162.15 | |
Powder |
4498-67-3 | |
Brown | |
MFCD00211066 | |
6354 | |
BHXVYTQDWMQVBI-UHFFFAOYSA-N | |
1H-indazole-3-carboxylic acid | |
78250 | |
98% |
Chemical Identifiers
4498-67-3 | |
162.148 | |
BHXVYTQDWMQVBI-UHFFFAOYSA-N | |
78250 | |
C1=CC=C2C(=C1)C(=NN2)C(=O)O |
C8H6N2O2 | |
MFCD00211066 | |
indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid | |
1H-indazole-3-carboxylic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-794-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only