missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H-Indazole-3-carboxylic acid, 98%

Manufacturer:  Alfa Aesar™ H6400122

 View more versions of this product

Catalog No. AAH6400122



Specifications

Specifications

1H-Indazole-3-carboxylic acid
266°C to 270°C (literature)
4498-67-3
MFCD00211066
indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
C1=CC=C2C(=C1)C(=NN2)C(=O)O
162.148
162.15
98%
Brown
100g
C8H6N2O2
6354
BHXVYTQDWMQVBI-UHFFFAOYSA-N
1H-indazole-3-carboxylic acid
78250
Powder

Chemical Identifiers

4498-67-3
162.148
BHXVYTQDWMQVBI-UHFFFAOYSA-N
78250
C1=CC=C2C(=C1)C(=NN2)C(=O)O
C8H6N2O2
MFCD00211066
indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
1H-indazole-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents