missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H-Imidazole-4,5-dicarboxylic Acid 98.0+%, TCI America™
$101.21 - $101.21
Chemical Identifiers
CAS | 570-22-9 |
---|---|
Molecular Formula | C5H4N2O4 |
Molecular Weight (g/mol) | 156.10 |
MDL Number | MFCD00005200 |
InChI Key | ZEVWQFWTGHFIDH-UHFFFAOYSA-N |
Synonym | 4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid |
PubChem CID | 68442 |
IUPAC Name | 1H-imidazole-4,5-dicarboxylic acid |
SMILES | OC(=O)C1=C(N=CN1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
I00035G
|
TCI America
I00035G |
5 g |
Each for $101.21
|
|
|||||
Chemical Identifiers
570-22-9 | |
156.10 | |
ZEVWQFWTGHFIDH-UHFFFAOYSA-N | |
68442 | |
OC(=O)C1=C(N=CN1)C(O)=O |
C5H4N2O4 | |
MFCD00005200 | |
4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid | |
1H-imidazole-4,5-dicarboxylic acid |
Specifications
570-22-9 | |
White-Yellow | |
MFCD00005200 | |
4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid | |
OC(=O)C1=C(N=CN1)C(O)=O | |
156.10 | |
156.10 | |
Crystalline Powder |
283°C | |
C5H4N2O4 | |
5 g | |
ZEVWQFWTGHFIDH-UHFFFAOYSA-N | |
1H-imidazole-4,5-dicarboxylic acid | |
68442 | |
≥98.0% (HPLC,T) | |
1H-Imidazole-4,5-dicarboxylic Acid |
Safety and Handling
EINECSNumber : (5)-6853
RTECSNumber : NI4760000
TSCA : Yes