missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Imidazole-2-carbaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 10111-08-7
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003544
InChI Key XYHKNCXZYYTLRG-UHFFFAOYSA-N
Synonym 1h-imidazole-2-carboxaldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, 2-formyl-1h-imidazole, 2-formylimidazole, 2-imidazolecarboxaldehyde, imidazol-2-carbaldehyde, imidazole-2-aldehyde, imidazole-2-carbaldehyde, imidazole-2-carboxaldehyde
PubChem CID 24955
IUPAC Name 1H-imidazole-2-carbaldehyde
SMILES O=CC1=NC=CN1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC08904DA
SDS View Documents Promotion Details
Fisher Scientific Company
CC08904DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10111-08-7
96.09
XYHKNCXZYYTLRG-UHFFFAOYSA-N
24955
O=CC1=NC=CN1
C4H4N2O
MFCD00003544
1h-imidazole-2-carboxaldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, 2-formyl-1h-imidazole, 2-formylimidazole, 2-imidazolecarboxaldehyde, imidazol-2-carbaldehyde, imidazole-2-aldehyde, imidazole-2-carbaldehyde, imidazole-2-carboxaldehyde
1H-imidazole-2-carbaldehyde

Specifications

Amber Glass Bottle
10111-08-7
100.0
C4H4N2O
1h-imidazole-2-carboxaldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, 2-formyl-1h-imidazole, 2-formylimidazole, 2-imidazolecarboxaldehyde, imidazol-2-carbaldehyde, imidazole-2-aldehyde, imidazole-2-carbaldehyde, imidazole-2-carboxaldehyde
O=CC1=NC=CN1
96.09
96.09
1g
94.5
95%
MFCD00003544
XYHKNCXZYYTLRG-UHFFFAOYSA-N
1H-imidazole-2-carbaldehyde
24955
1H-imidazole-2-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Product Certifications