missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Imidazol-2-ylmethanol hydrochloride, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC08909DE

 View more versions of this product

Catalog No. CC08909DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

134.57
97%
116177-22-1
MFCD03659700
C4H6N2O·ClH
1H-imidazol-2-ylmethanol hydrochloride
5g
100.0
96.5
Amber Glass Bottle
97%

Chemical Identifiers

116177-22-1
100.0
C4H6N2O·ClH
MFCD03659700
97%
96.5
97%
134.57
1H-imidazol-2-ylmethanol hydrochloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.