missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Benzotriazole, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1542318

 View more versions of this product

Catalog No. AAA1542318



Description

Description

Applications
1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.

Solubility
Soluble in water, benzene, toluene, chloroform, ethanol and N,N-dimethylformamide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and heavy metals.
Specifications

Specifications

Benzotriazole
50g
99%
MFCD00005699
14,1108
QRUDEWIWKLJBPS-UHFFFAOYSA-N
2H-benzotriazole
7220
119.13
1.360
95-14-7
C6H5N3
112133
1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
C1=CC2=NNN=C2C=C1
119.127
CHEBI:75331
99%

Chemical Identifiers

95-14-7
119.127
QRUDEWIWKLJBPS-UHFFFAOYSA-N
7220
2H-benzotriazole
C6H5N3
MFCD00005699
1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
CHEBI:75331
C1=CC2=NNN=C2C=C1
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H319

GHS P Statement
P273-P280h-P305+P351+P338

Warning

RTECSNumber : DM1225000

TSCA : TSCA

Documents