missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H-Benzimidazole-5-carbaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC45904DA
Specifications
1H-Benzimidazole-5-carbaldehyde | |
97% | |
C8H6N2O | |
1h-benzimidazole-5-carbaldehyde, 1h-benzimidazole-5-carboxaldehyde, 1h-benzo d imidazole-5-carbaldehyde, benzimidazole-5-aldehyde, 1h-1,3-benzodiazole-5-carbaldehyde, 1h-benzimidazole-6-carboxaldehyde, benzimidazole-5-carboxaldehyde, 1h-benzoimidazole-5-carbaldehyde, 3h-benzoimidazole-5-carbaldehyde, pubchem9866 | |
C1=CC2=C(C=C1C=O)NC=N2 | |
146.149 | |
146.15 |
58442-17-4 | |
Amber Glass Bottle | |
1g | |
ALCHVVTYAHQOFY-UHFFFAOYSA-N | |
3H-benzimidazole-5-carbaldehyde | |
10012038 | |
97% |
Chemical Identifiers
58442-17-4 | |
146.149 | |
1h-benzimidazole-5-carbaldehyde, 1h-benzimidazole-5-carboxaldehyde, 1h-benzo d imidazole-5-carbaldehyde, benzimidazole-5-aldehyde, 1h-1,3-benzodiazole-5-carbaldehyde, 1h-benzimidazole-6-carboxaldehyde, benzimidazole-5-carboxaldehyde, 1h-benzoimidazole-5-carbaldehyde, 3h-benzoimidazole-5-carbaldehyde, pubchem9866 | |
3H-benzimidazole-5-carbaldehyde |
C8H6N2O | |
ALCHVVTYAHQOFY-UHFFFAOYSA-N | |
10012038 | |
C1=CC2=C(C=C1C=O)NC=N2 |