Please login to your online account to display your discounted pricing

1H-Benzimidazole-5-carbaldehyde, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC45904CB

 View more versions of this product

Catalog No. CC45904CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 146.15
Quantity 250mg
Assay 97%
Chemical Name or Material 1H-Benzimidazole-5-carbaldehyde
CAS 58442-17-4
Molecular Formula C8H6N2O
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 1H-Benzimidazole-5-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6N2O
Formula Weight 146.15