missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Benzimidazole-4-carboxylic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC429510010

 View more versions of this product

Catalog No. AC429510010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1H-Benzimidazole-4-carboxylicacid
1g
96.0
96% min. (HPLC)
MFCD01823426
VVQNAFBGAWCMLU-UHFFFAOYSA-N
1H-benzimidazole-4-carboxylic acid
2771758
97%
Glass bottle
46006-36-4
100.0
C8H6N2O2
1h-1,3-benzodiazole-4-carboxylic acid, 1h-benzimidazole-4-carboxylicacid, 1h-benzimidazole-7-carboxylic acid, 1h-benzo d imidazole-4-carboxylic acid, 1h-benzo d imidazole-7-carboxylic acid, 1h-benzoimidazole-4-carboxylic acid, 3h-1,3-benzodiazole-4-carboxylic acid, 4-carboxy-1h-benzimidazole, acmc-1aqhf, benzimidazole-4-carboxylic acid
C1=CC(=C2C(=C1)NC=N2)C(=O)O
162.15
162.15

Chemical Identifiers

46006-36-4
162.15
VVQNAFBGAWCMLU-UHFFFAOYSA-N
2771758
C1=CC(=C2C(=C1)NC=N2)C(=O)O
C8H6N2O2
MFCD01823426
1h-1,3-benzodiazole-4-carboxylic acid, 1h-benzimidazole-4-carboxylicacid, 1h-benzimidazole-7-carboxylic acid, 1h-benzo d imidazole-4-carboxylic acid, 1h-benzo d imidazole-7-carboxylic acid, 1h-benzoimidazole-4-carboxylic acid, 3h-1,3-benzodiazole-4-carboxylic acid, 4-carboxy-1h-benzimidazole, acmc-1aqhf, benzimidazole-4-carboxylic acid
1H-benzimidazole-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents