Please login to your online account to display your discounted pricing

1H-Benzimidazole-2-methanol, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  304190250

 View more versions of this product

Catalog No. AC304190250

  • / Each
 

Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 170° to 175°C
Color Off-white to light brown
Quantity 25g
Chemical Name or Material 1H-Benzimidazole-2-methanol
Name Note 97%
CAS 4856-97-7
CAS Min % 96.0
CAS Max % 100.0
Physical Form Off-white to light brown crystalline powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H8N2O
Formula Weight 148.16g/mol
MDL Number MFCD00014560
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 225-451-2
TSCA TSCA

Specifications

Chemical Name or Material 1H-Benzimidazole-2-methanol
Name Note 97%
CAS 4856-97-7
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H8N2O
Formula Weight 148.16g/mol
MDL Number MFCD00014560
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents