missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Benzimidazole-2-methanol, 97%, ACROS Organics™

Chemical Identifiers

CAS 4856-97-7
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
MDL Number MFCD00014560
InChI Key IAJLTMBBAVVMQO-UHFFFAOYSA-N
Synonym 1h-1,3-benzodiazol-2-ylmethanol, 1h-benzimidazol-2-yl methanol, 1h-benzimidazole-2-methanol, 1h-benzo d imidazol-2-yl methanol, 1h-benzoimidazol-2-yl methanol, 1h-benzoimidazol-2-yl-methanol, 2-benzimidazolemethanol, 2-hydroxymethyl benzimidazole, 2-hydroxymethyl-1h-benzimidazole, 2-hydroxymethylbenzimidazole
PubChem CID 78569
IUPAC Name 1H-benzimidazol-2-ylmethanol
SMILES C1=CC=C2C(=C1)NC(=N2)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC304190010
SDS View Documents
Thermo Fisher Scientific
304190010
1g Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

4856-97-7
148.165
IAJLTMBBAVVMQO-UHFFFAOYSA-N
78569
C1=CC=C2C(=C1)NC(=N2)CO
C8H8N2O
MFCD00014560
1h-1,3-benzodiazol-2-ylmethanol, 1h-benzimidazol-2-yl methanol, 1h-benzimidazole-2-methanol, 1h-benzo d imidazol-2-yl methanol, 1h-benzoimidazol-2-yl methanol, 1h-benzoimidazol-2-yl-methanol, 2-benzimidazolemethanol, 2-hydroxymethyl benzimidazole, 2-hydroxymethyl-1h-benzimidazole, 2-hydroxymethylbenzimidazole
1H-benzimidazol-2-ylmethanol

Specifications

Authentic
White to Brown
1g
96.0
97%
MFCD00014560
IAJLTMBBAVVMQO-UHFFFAOYSA-N
1H-benzimidazol-2-ylmethanol
78569
Crystalline Powder
1H-Benzimidazole-2-methanol, 97%
Glass Bottle
170°C to 175°C
4856-97-7
100.0
C8H8N2O
1h-1,3-benzodiazol-2-ylmethanol, 1h-benzimidazol-2-yl methanol, 1h-benzimidazole-2-methanol, 1h-benzo d imidazol-2-yl methanol, 1h-benzoimidazol-2-yl methanol, 1h-benzoimidazol-2-yl-methanol, 2-benzimidazolemethanol, 2-hydroxymethyl benzimidazole, 2-hydroxymethyl-1h-benzimidazole, 2-hydroxymethylbenzimidazole
C1=CC=C2C(=C1)NC(=N2)CO
148.165
148.16
≥96.0%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 225-451-2

TSCA : TSCA