missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H-Benzimidazole-2-carboxylic acid hydrate, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC09051CB
Specifications
1H-Benzimidazole-2-carboxylic acid hydrate | |
90% | |
C8H8N2O3 | |
1h-benzo d imidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid monohydrate, benzimidazole-2-carboxylic acid monohydrate, 1h-1,3-benzodiazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylicacid, hydrate 1:1, c8h6n2o2.h2o, 2-carboxy-1h-benzimidazole hydrate, 1h-benzimidazole-2-carboxylic acid-water 1/1 | |
C1=CC=C2C(=C1)NC(=N2)C(=O)O.O | |
180.163 | |
180.17 |
849776-47-2 | |
Amber Glass Bottle | |
250mg | |
YQHUUYLPOCNBKD-UHFFFAOYSA-N | |
1H-benzimidazole-2-carboxylic acid;hydrate | |
18525703 | |
90% |
Chemical Identifiers
849776-47-2 | |
180.163 | |
1h-benzo d imidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid monohydrate, benzimidazole-2-carboxylic acid monohydrate, 1h-1,3-benzodiazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylicacid, hydrate 1:1, c8h6n2o2.h2o, 2-carboxy-1h-benzimidazole hydrate, 1h-benzimidazole-2-carboxylic acid-water 1/1 | |
1H-benzimidazole-2-carboxylic acid;hydrate |
C8H8N2O3 | |
YQHUUYLPOCNBKD-UHFFFAOYSA-N | |
18525703 | |
C1=CC=C2C(=C1)NC(=N2)C(=O)O.O |
Safety and Handling
Warning