missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Benzimidazol-5-ylmethanol, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC45909DA

 View more versions of this product

Catalog No. CC45909DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

148.16
95%
95%
Amber Glass Bottle
MFCD08690267
C8H8N2O
1g
100.0
106429-29-2
1H-benzimidazol-5-ylmethanol
94.5

Chemical Identifiers

106429-29-2
100.0
C8H8N2O
MFCD08690267
95%
94.5
95%
148.16
1H-benzimidazol-5-ylmethanol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning