missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Benzimidazol-5-ylmethanol, 97%, Maybridge

$64.23 - $64.23

Chemical Identifiers

CAS 106429-29-2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8H8N2O
Formula Weight 148.16
MDL Number MFCD08690267
Chemical Name or Material 1H-benzimidazol-5-ylmethanol
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC45909CB SDS Fisher Scientific Uk Ltd
CC45909CB
Amber Glass Bottle 250mg Each for $64.23
CC45909DA SDS Fisher Scientific Company
CC45909DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

106429-29-2
100.0
C8H8N2O
MFCD08690267
95%
94.5
95%
148.16
1H-benzimidazol-5-ylmethanol

Specifications

106429-29-2
100.0
95%
148.16
250mg
95%
94.5
Amber Glass Bottle
C8H8N2O
MFCD08690267
1H-benzimidazol-5-ylmethanol

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning