Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H-Perfluorooctyl methacrylate, 97%

$45.65 - $140.88

Chemical Identifiers

Chemical Name or Material 1H,1H-Perfluorooctyl methacrylate
CAS 3934-23-4
Molecular Formula C12H7F15O2
Formula Weight 468.16
MDL Number MFCD00039247
Beilstein 6445701
Synonym Methacrylic acid 1H,1H-perfluorooctyl ester; 1H,1H-Pentadecafluorooctyl methacrylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1044003

  SDS
alfa aesar™
L1044003
1g Each for $45.65

AAL1044006

  SDS
alfa aesar™
L1044006
5g Each for $140.88
Description & Specifications

Specifications

Chemical Name or Material 1H,1H-Perfluorooctyl methacrylate
Flash Point 96°C (204°F)
Boiling Point 70°C (1mmHg)
Quantity 1g
CAS 3934-23-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
TSCA TSCA
Assay 97%
Molecular Formula C12H7F15O2
Formula Weight 468.16
MDL Number MFCD00039247
Beilstein 6445701
Synonym Methacrylic acid 1H,1H-perfluorooctyl ester; 1H,1H-Pentadecafluorooctyl methacrylate
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C