missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H-Perfluorooctyl methacrylate, 97%

Manufacturer:  Alfa Aesar™ L1044006

 View more versions of this product

Catalog No. AAL1044006



Specifications

Specifications

1H,1H-Perfluorooctyl methacrylate
70°C (1mmHg)
3934-23-4
MFCD00039247
1h,1h-perfluorooctyl methacrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, 1h,1h-perfluoro-n-octyl methacrylate, 1h,1h-perfluorooctyl methacrylate, contains mehq as inhibitor, 2-propenoic acid,2-methyl-, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, methacrylic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester
CC(=C)C(=O)OCC(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
468.162
468.16
96°C (204°F)
5g
C12H7F15O2
6445701
RUEKTOVLVIXOHT-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl 2-methylprop-2-enoate
77532
97%

Chemical Identifiers

3934-23-4
468.162
RUEKTOVLVIXOHT-UHFFFAOYSA-N
77532
CC(=C)C(=O)OCC(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C12H7F15O2
MFCD00039247
1h,1h-perfluorooctyl methacrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, 1h,1h-perfluoro-n-octyl methacrylate, 1h,1h-perfluorooctyl methacrylate, contains mehq as inhibitor, 2-propenoic acid,2-methyl-, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, methacrylic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents