missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H-Perfluorooctyl acrylate, 97%, stab. with 50ppm 4-methoxyphenol

$56.09 - $448.77

Chemical Identifiers

CAS 307-98-2
Molecular Formula C11F15H5O2
Linear Formula CF3(CF2)6CH2O2CCH=CH2
Formula Weight 454.14
MDL Number MFCD00039248
Beilstein 1809801
Synonym Acrylic acid 1H,1H-perfluorooctyl ester; 1H,1H-Pentadecafluorooctyl acrylate
Chemical Name or Material 1H,1H-Perfluorooctyl acrylate, stabilized with 50ppm 4-methoxyphenol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1660303 SDS Alfa Aesar™
L1660303
1g Each for $56.09
AAL1660306 SDS Alfa Aesar™
L1660306
5g Each for $120.86
AAL1660314 SDS Alfa Aesar™
L1660314
25g Each for $448.77
Specifications Description & Specifications

Chemical Identifiers

307-98-2
CF3(CF2)6CH2O2CCH=CH2
MFCD00039248
Acrylic acid 1H,1H-perfluorooctyl ester; 1H,1H-Pentadecafluorooctyl acrylate
C11F15H5O2
454.14
1809801
1H,1H-Perfluorooctyl acrylate, stabilized with 50ppm 4-methoxyphenol

Specifications

91°C (195°F)
1g
C11F15H5O2
454.14
1809801
1H,1H-Perfluorooctyl acrylate, stabilized with 50ppm 4-methoxyphenol
112° to 115°C (60mmHg)
307-98-2
CF3(CF2)6CH2O2CCH=CH2
MFCD00039248
Acrylic acid 1H,1H-perfluorooctyl ester; 1H,1H-Pentadecafluorooctyl acrylate
97%

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C