Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H-Perfluoro-1-tetradecanol, 96%

$102.05 - $354.04

Chemical Identifiers

Chemical Name or Material 1H,1H-Perfluoro-1-tetradecanol
CAS 15622-57-8
Molecular Formula CF3(CF2)12CH2OH
Formula Weight 700.14
MDL Number MFCD00153264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1660914

  SDS
alfa aesar™
L1660914
25g Each for $354.04

AAL1660906

  SDS
alfa aesar™
L1660906
5g Each for $102.05
Description & Specifications

Specifications

Formula Weight 700.14
Chemical Name or Material 1H,1H-Perfluoro-1-tetradecanol
Molecular Formula CF3(CF2)12CH2OH
MDL Number MFCD00153264
CAS 15622-57-8
Assay 96%
Boiling Point 130°C (60mmHg)
Melting Point 137° to 140°C
Quantity 25g