missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H,1H-Perfluoro-1-octanol, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 277260050
Specifications
1H, 1H-Perfluoro-1-octanol | |
Authentic | |
100°C to 102°C (100.0 hPa) | |
43.0°C to 47.0°C | |
307-30-2 | |
100.0 | |
C8H3F15O | |
MFCD00004675 | |
1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, 1h,1h-pentadecafluoro-1-octanol, 1h,1h-perfluoro-1-octanol, 1h,1h-perfluorooctanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl alcohol, pentadecafluoro-1-octanol, pentadecafluorooctyl alcohol, perfluorooctanol, unii-9x5w7fq62r | |
PJDOLCGOTSNFJM-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol | |
67543 | |
Crystalline Mass |
>150°C | |
Glass bottle | |
White | |
5g | |
97.5 | |
97.5% min. (GC) | |
CF3(CF2)6CH2OH | |
01, IV, 1766 | |
Solubility in water: insoluble | |
C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
400.08 | |
400.08 | |
98% |
Chemical Identifiers
307-30-2 | |
400.08 | |
PJDOLCGOTSNFJM-UHFFFAOYSA-N | |
67543 | |
C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
C8H3F15O | |
MFCD00004675 | |
1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, 1h,1h-pentadecafluoro-1-octanol, 1h,1h-perfluoro-1-octanol, 1h,1h-perfluorooctanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl alcohol, pentadecafluoro-1-octanol, pentadecafluorooctyl alcohol, perfluorooctanol, unii-9x5w7fq62r | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 206-197-1