Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H-Perfluoro-1-octanol, 97%

$67.05 - $200.41

Chemical Identifiers

Chemical Name or Material 1H,1H-Perfluoro-1-octanol
CAS 307-30-2
Molecular Formula CF3(CF2)6CH2OH
Formula Weight 400.08
MDL Number MFCD00004675
Beilstein 1716494
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluoro-1-octanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1803314

  SDS
alfa aesar™
A1803314
25g Each for $200.41

AAA1803306

  SDS
alfa aesar™
A1803306
5g Each for $67.05
Description & Specifications

Specifications

Formula Weight 400.08
Assay 97%
Molecular Formula CF3(CF2)6CH2OH
MDL Number MFCD00004675
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluoro-1-octanol
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
CAS 307-30-2
Chemical Name or Material 1H,1H-Perfluoro-1-octanol
Melting Point 43° to 47°C
Quantity 25g
Boiling Point 160° to 162°C
Beilstein 1716494
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.