Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H-Perfluoro-1-nonanol, 98%

$75.19 - $75.19

Chemical Identifiers

Chemical Name or Material 1H,1H-Perfluoro-1-nonanol
CAS 423-56-3
Molecular Formula C9H3F17O
Linear Formula CF3(CF2)7CH2OH
Formula Weight 450.1
MDL Number MFCD00153183
Beilstein 1809099
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-nonanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1660006

  SDS
alfa aesar™
L1660006
5g Each for $75.19
Description & Specifications

Specifications

Formula Weight 450.1
Chemical Name or Material 1H,1H-Perfluoro-1-nonanol
CAS 423-56-3
MDL Number MFCD00153183
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-nonanol
Molecular Formula C9H3F17O
Assay 98%
Linear Formula CF3(CF2)7CH2OH
Beilstein 1809099
Quantity 5g
Melting Point 66° to 71°C
Boiling Point 176°C

Applications
1H,1H-Perfluoro-1-nonanol is used to produce other chemicals. For example, it is used to produce Trifluoro-methanesulfonic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-nonyl ester. This reaction needs reagent Pyridine and solvent CHCl3

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.