missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H,1H-Pentadecafluoro-n-octyl Acrylate (stabilized with MEHQ), TCI America™

Manufacturer:  TCI America P17541G

 View more versions of this product

Catalog No. P17541G



Specifications

Specifications

1H,1H-Pentadecafluoro-n-octyl Acrylate (stabilized with MEHQ)
Yellow
307-98-2
MFCD00039248
YSQGYEYXKXGAQA-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl prop-2-enoate
67551
Clear Liquid at 20°C
65°C
1g
C11H5F15O2
1h,1h-pentadecafluorooctyl acrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl acrylate, 1h,1h-perfluorooctyl acrylate, acrylic acid 1h,1h-pentadecafluoro-n-octyl ester, 2-propenoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, 1h,1h-perfluoro-n-octyl acrylate, 1h,1h-pentadecafluoro-n-octyl acrylate, 1,1-dihydroperfluorooctyl acrylate, 1,1-dyhydroperfluorooctyl acrylate, acrylic acid 1h,1h-perfluorooctyl ester
C=CC(=O)OCC(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
454.135
454.14
≥95.0% (GC)

Chemical Identifiers

307-98-2
454.135
YSQGYEYXKXGAQA-UHFFFAOYSA-N
67551
C=CC(=O)OCC(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C11H5F15O2
MFCD00039248
1h,1h-pentadecafluorooctyl acrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl acrylate, 1h,1h-perfluorooctyl acrylate, acrylic acid 1h,1h-pentadecafluoro-n-octyl ester, 2-propenoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, 1h,1h-perfluoro-n-octyl acrylate, 1h,1h-pentadecafluoro-n-octyl acrylate, 1,1-dihydroperfluorooctyl acrylate, 1,1-dyhydroperfluorooctyl acrylate, acrylic acid 1h,1h-perfluorooctyl ester
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl prop-2-enoate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents