missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,9H,9H-Perfluoro-1,9-nonanediol, tech. 90%

$94.53 - $271.42

Chemical Identifiers

CAS 203303-01-9
Molecular Formula C9H6F14O2
Molecular Weight (g/mol) 412.123
MDL Number MFCD00153275
InChI Key XQULMKMNFZLURS-UHFFFAOYSA-N
Synonym 1h,1h,9h,9h-perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro-1,9-nonanediol, 1h,1h,9h,9h-perfluorononane-1,9-diol, 1h,1h,9h,9h-perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonane-1,9-diol
PubChem CID 2776243
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononane-1,9-diol
SMILES C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1659906 View Documents Alfa Aesar™
L1659906
5g Each for $94.53
 
AAL1659914 View Documents Alfa Aesar™
L1659914
25g Each for $271.42
 
Specifications

Chemical Identifiers

203303-01-9
412.123
XQULMKMNFZLURS-UHFFFAOYSA-N
2776243
C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
C9H6F14O2
MFCD00153275
1h,1h,9h,9h-perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro-1,9-nonanediol, 1h,1h,9h,9h-perfluorononane-1,9-diol, 1h,1h,9h,9h-perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonane-1,9-diol
2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononane-1,9-diol

Specifications

140°C (5mmHg)
5g
C9H6F14O2
MFCD00153275
XQULMKMNFZLURS-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononane-1,9-diol
2776243
90%
1H,1H,9H,9H-Perfluoro-1,9-nonanediol
108°C to 112°C
203303-01-9
HOCH2(CF2)7CH2OH
1h,1h,9h,9h-perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro-1,9-nonanediol, 1h,1h,9h,9h-perfluorononane-1,9-diol, 1h,1h,9h,9h-perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonane-1,9-diol
C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
412.123
412.12
Technical