missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,8H,8H-Dodecafluoro-1,8-octanediol, 97%

$43.03 - $43.03

Chemical Identifiers

CAS 90177-96-1
Molecular Formula C8H6F12O2
Molecular Weight (g/mol) 362.115
MDL Number MFCD00153145
InChI Key XZJPYETUABEQFI-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-perfluorooctane-1,8-diol, 1,8-dihydroxy-2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane, 1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, 1h,1h,8h,8h-perfluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluorooctane-1,8-diol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octane-1,8-diol
PubChem CID 3820533
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane-1,8-diol
SMILES C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1648403 View Documents Alfa Aesar™
L1648403
1g Each for $43.03
 
Description

Description

Applications
1H,1H,8H,8H-Dodecafluoro-1,8-octanediol is used in pharma industry.

Solubility
Soluble in methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

90177-96-1
362.115
XZJPYETUABEQFI-UHFFFAOYSA-N
3820533
C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
C8H6F12O2
MFCD00153145
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-perfluorooctane-1,8-diol, 1,8-dihydroxy-2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane, 1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, 1h,1h,8h,8h-perfluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluorooctane-1,8-diol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octane-1,8-diol
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane-1,8-diol

Specifications

105°C (0.08mmHg)
1g
C8H6F12O2
MFCD00153145
XZJPYETUABEQFI-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane-1,8-diol
3820533
97%
80°C to 88°C
90177-96-1
HOCH2(CF2)6CH2OH
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-perfluorooctane-1,8-diol, 1,8-dihydroxy-2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane, 1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, 1h,1h,8h,8h-perfluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluorooctane-1,8-diol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octane-1,8-diol
C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
362.115
362.12
1H,1H,8H,8H-Dodecafluoro-1,8-octanediol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning