Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,7H-Dodecafluoroheptyl acrylate, 97%, stab. with 50ppm 4-methoxyphenol

Chemical Identifiers

Chemical Name or Material 1H,1H,7H-Dodecafluoroheptyl acrylate
CAS 2993-85-3
Name Note stabilized with 50ppm 4-methoxyphenol
Molecular Formula C10H6F12O2
Linear Formula CH2=CHCO2CH2(CF2)6H
Formula Weight 386.14
MDL Number MFCD00080746
UN Number UN3082
Beilstein 3563773
Synonym Acrylic acid 1H,1H,7H-dodecafluoroheptyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1648206

  SDS
alfa aesar™
L1648206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1648214

  SDS
alfa aesar™
L1648214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 386.14
Assay 97%
Molecular Formula C10H6F12O2
Beilstein 3563773
Synonym Acrylic acid 1H,1H,7H-dodecafluoroheptyl ester
MDL Number MFCD00080746
UN Number UN3082
Name Note stabilized with 50ppm 4-methoxyphenol
Flash Point 101°C (213°F)
Recommended Storage 2° to 8°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P273-P305+P351+P338-P302+P352
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Health Hazard 1 Warning
Chemical Name or Material 1H,1H,7H-Dodecafluoroheptyl acrylate
RTECS Number UD3340500
Linear Formula CH2=CHCO2CH2(CF2)6H
Quantity 5g
Odor Pungent
Boiling Point 197°C
CAS 2993-85-3
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P273-P305+P351+P338-P302+P352
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C