Please login to your online account to display your discounted pricing

1H,1H,7H-Dodecafluoroheptanol, 97%, Acros Organics


Quantity 5mL

Manufacturer: thermo fisher scientific  336070050

 View more versions of this product

Catalog No. AC336070050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 332.09
Physical Form Liquid
Quantity 5mL
Color Clear colorless
TSCA TSCA
EINECS Number 206-406-6
Density 1.7600g/mL
CAS Min % 96.0
CAS Max % 100.0
Synonym 1H, 1H, 7H-Perfluoro-1-heptanol
Chemical Name or Material 1H, 1H, 7H-Dodecafluoroheptanol
Refractive Index 1.3160 to 1.3180 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Linear Formula CHF2(CF2)5CH2OH
Name Note 97%
MDL Number MFCD00039630
Assay Percent Range 97%
Molecular Formula C7H4F12O
CAS 335-99-9

Specifications

Chemical Name or Material 1H, 1H, 7H-Dodecafluoroheptanol
Name Note 97%
CAS 335-99-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4F12O
Linear Formula CHF2(CF2)5CH2OH
Formula Weight 332.09
MDL Number MFCD00039630
Synonym 1H, 1H, 7H-Perfluoro-1-heptanol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents