missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H,1H,5H-Octafluoropentyl Methacrylate (stabilized with TBC), TCI America™

$64.17 - $186.39

Chemical Identifiers

CAS 355-93-1
Molecular Formula C9H8F8O2
Molecular Weight (g/mol) 300.148
MDL Number MFCD00039278
InChI Key ZNJXRXXJPIFFAO-UHFFFAOYSA-N
Synonym 1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate
PubChem CID 67739
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(C(C(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14335G View Documents TCI America
M14335G
5g Each for $64.17
 
M143325G View Documents TCI America
M143325G
25g Each for $186.39
 
Specifications

Chemical Identifiers

355-93-1
300.148
ZNJXRXXJPIFFAO-UHFFFAOYSA-N
67739
CC(=C)C(=O)OCC(C(C(C(F)F)(F)F)(F)F)(F)F
C9H8F8O2
MFCD00039278
1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate
2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate

Specifications

88°C
5g
C9H8F8O2
1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate
CC(=C)C(=O)OCC(C(C(C(F)F)(F)F)(F)F)(F)F
300.148
300.15
≥98.0% (GC)
Yellow
355-93-1
MFCD00039278
ZNJXRXXJPIFFAO-UHFFFAOYSA-N
2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate
67739
Clear Liquid at 20°C
1H,1H,5H-Octafluoropentyl Methacrylate (stabilized with TBC)
Safety and Handling

Safety and Handling

TSCA : No